Learn More
Cayman Chemical MPP+ Iodide, Purity: ≥98%, CAS: 36913-39-0, 50 mg
SDP
Catalog No. 502966146
Encompass_Preferred
An active metabolite of MPTP, a neurotoxin used to cause selective destruction of dopaminergic neurons in animal models of parkinsonism; inhibits complex I of the mitochondrial electron transport chain, resulting in ATP depletion and increased oxidative stress
Catalog No. 50-296-6146 Supplier Cayman Chemical Supplier No. 1695850 MG
An active metabolite of MPTP, a neurotoxin used to cause selective destruction of dopaminergic neurons in animal models of parkinsonism; inhibits complex I of the mitochondrial electron transport chain, resulting in ATP depletion and increased oxidative stress