Learn More
Selleck Chemical LLC MPP iodide
SDP
Supplier:  Selleck Chemical LLC S63011G
Encompass_Preferred
MPP iodide (N-Methyl-4-Phenylpyridinium Iodide) the metabolite of a neurotoxin MPTP causes symptom of Parkinson s disease (PD) in animal models by selectively destroying dopaminergic neurons in substantia nigra MPP induces autophagic cell death in SH-SY5Y cells MPP induces dopamine transporter (DAT) externalization in dopaminergic (DA) neurons but internalization of serotonin transporter (SERT) in serotonergic (5-HT) neurons
Catalog No. 50-000-03123