Learn More
Aviva Systems Biology Corp MPP2 Antibody - middle region (ARP56632_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56632_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MPP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MPP2. Alternate names: DKFZp686A06252, DKFZp686J2189, DKFZp761D0712, DLG2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61933