Learn More
Aviva Systems Biology Corp MPP4 Antibody - C-terminal region (ARP65025_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65025_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MPP4. It was validated on Western Blot. . Alternate names: ALS2CR5, DLG6ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-288-415