Learn More
Aviva Systems Biology Corp MPP7 Antibody - N-terminal region (ARP53247_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53247_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MPP7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MPP7. Alternate names: FLJ32798, RP11-218D6.5ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60982