Learn More
Cayman Chemical MPPodde 100mg
SDP
Catalog No. 502980019
Encompass_Preferred
An active metabolite of MPTP, a neurotoxin used to cause selective destruction of dopaminergic neurons in animal models of parkinsonism; inhibits complex I of the mitochondrial electron transport chain, resulting in ATP depletion and increased oxidative stress
Catalog No. 50-298-0019 Supplier Cayman Chemical Supplier No. 16958100 MG
An active metabolite of MPTP, a neurotoxin used to cause selective destruction of dopaminergic neurons in animal models of parkinsonism; inhibits complex I of the mitochondrial electron transport chain, resulting in ATP depletion and increased oxidative stress