Learn More
Electron Microscopy Sciences mPrep/f™ 13 Extreme filter-couplers, storage box
SDP
Supplier:  Electron Microscopy Sciences 8501012
Encompass_Preferred
Filter couplers prevent the introduction of damaging reagents into pipettors. They also improve the fit of mPrep/g capsules on some pipettors. Pack includes 16 filter couplers and a capsule storage box. 13 µm pore size filter for use with biohazards and very aggressive reagents.
Catalog No. 50-192-8120