Learn More
Electron Microscopy Sciences mPrep™ Pipettor Kit Single channel
SDP
Catalog No. 501928116
Encompass_Preferred
Pipettor kits for use with mPrep/s™ or mPrep/g™ capsules. Single-Channel Kit includes: single channel 200 microliter pipettor, one mPrep/g capsule pack and one pack of 96 pipette tips, size 10µl.
Catalog No. 50-192-8116 Supplier Electron Microscopy Sciences Supplier No. 8501007
Pipettor kits for use with mPrep/s™ or mPrep/g™ capsules. Single-Channel Kit includes: single channel 200 microliter pipettor, one mPrep/g capsule pack and one pack of 96 pipette tips, size 10µl.