Learn More
Electron Microscopy Sciences mPrep™ Pipettor Kit Multi-channel
SDP
Catalog No. 501928117
Encompass_Preferred
Pipettor kits for use with mPrep/s™ or mPrep/g™ capsules. Eight-Channel Kit includes: eight-channel 200 microliter pipettor, one mPrep/g capsule pack and one pack of 96 pipette tips, size 10µl.
Catalog No. 50-192-8117 Supplier Electron Microscopy Sciences Supplier No. 8501008
Pipettor kits for use with mPrep/s™ or mPrep/g™ capsules. Eight-Channel Kit includes: eight-channel 200 microliter pipettor, one mPrep/g capsule pack and one pack of 96 pipette tips, size 10µl.