Learn More
Electron Microscopy Sciences mPrep/s™ Workstation, complete
SDP
Supplier:  Electron Microscopy Sciences 8501006
Encompass_Preferred
The mPrep/s WorkStation is for prepraing biological tissue for electron microscopy using the mPrep/s capsules. It can be used for capsules of tissue for TEM, fixing of tissue for critical point drying (CPD), bio tissue cryo-facing, and polymer cross-sections.
Simply load the capsule onto the built-in insertion tool at the center of the workstation, detach the screen from the capsule, use the lever to open the screen and insert the specimen. Then release the lever and re-attach the capsule. Once loaded, the sample requires no additional handling even for TEM embedding or SEM mounting.
The polyethylene surface is fully immersible for easy cleaning between uses. Molded into the surface are 12 dissection wells. At the back, 12 capsule wells and 3 screen holders conveniently hold these prior to use. On either side of the workstation, a total of 24 capsule wells can hold loaded capsules and keep them wet while loading into the channels of the pipettor.
Catalog No. 50-192-8115