Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mpro inhibitor N3 | 884650-98-0 | 99.8% | 680.79 | 1 ML
SDP

Catalog No. 5000350241
Encompass_Preferred

Mpro inhibitor N3 is a potent SARS-CoV-2 MPro inhibitor with an EC50 value of 16.77 μM. It also exhibits antiviral activities against HCoV-229E, FIPV, IBV, and MHV-A59.

  • Potent SARS-CoV-2 MPro inhibitor (EC50 16.77 μM).
  • Exhibits antiviral activity against HCoV-229E, FIPV, IBV, and MHV-A59.
  • Soluble in DMSO (20 mg/mL).
  • Store at 4°C, sealed, away from moisture and light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Appearance: Off-white to light yellow solid.

Catalog No. 50-003-50241 Supplier Medchemexpress LLC Supplier No. HY13614910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mpro inhibitor N3 is a potent SARS-CoV-2 MPro inhibitor with an EC50 value of 16.77 μM. It also exhibits antiviral activities against HCoV-229E, FIPV, IBV, and MHV-A59.

  • Potent SARS-CoV-2 MPro inhibitor (EC50 16.77 μM).
  • Exhibits antiviral activity against HCoV-229E, FIPV, IBV, and MHV-A59.
  • Soluble in DMSO (20 mg/mL).
  • Store at 4°C, sealed, away from moisture and light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Appearance: Off-white to light yellow solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.