Learn More
Medchemexpress LLC Mpro inhibitor N3 (hemihydrate) | 99.9% | 689.80 | 25 MG
SDP
Catalog No. 5000351586
Encompass_Preferred
This product is a potent inhibitor of SARS-CoV-2 Mpro with an EC50 of 16.77 μM. It specifically inhibits Mpro from various coronaviruses, including SARS-CoV and MERS-CoV, and demonstrates inhibitory activity against HCoV-229E, FIPV, and MHV-A59 with respective IC50 values of 4.0 μM, 8.8 μM, and 2.7 μM.
  • Potent inhibitor of SARS-CoV-2 Mpro.
  • Exhibits broad-spectrum inhibition against multiple coronaviruses.
  • Demonstrates inhibition against various HCoVs.
  • Able to penetrate cells to inhibit virus replication in embryos.
Catalog No. 50-003-51586 Supplier Medchemexpress LLC Supplier No. 25MGHY136149A
This product is a potent inhibitor of SARS-CoV-2 Mpro with an EC50 of 16.77 μM. It specifically inhibits Mpro from various coronaviruses, including SARS-CoV and MERS-CoV, and demonstrates inhibitory activity against HCoV-229E, FIPV, and MHV-A59 with respective IC50 values of 4.0 μM, 8.8 μM, and 2.7 μM.
  • Potent inhibitor of SARS-CoV-2 Mpro.
  • Exhibits broad-spectrum inhibition against multiple coronaviruses.
  • Demonstrates inhibition against various HCoVs.
  • Able to penetrate cells to inhibit virus replication in embryos.