Learn More
Medchemexpress LLC Mpro inhibitor N3 hemihydrate | MFCD00274570 | 99.9% | 698.81 | C35H50N6O9 | 5 MG
SDP
Catalog No. 5000222866
Encompass_Preferred
Mpro inhibitor N3 hemihydrate is a small-molecule inhibitor of coronavirus main protease (Mpro) provided as a white to off-white solid hemihydrate for biochemical and antiviral research; product page lists purity, storage, and solubility information for experimental planning.
  • Potent inhibitor of coronavirus main protease with reported cellular activity.
  • High purity (99.86%).
  • Supplied as a white to off-white solid hemihydrate.
  • Stable when stored as powder at -20°C for up to 3 years.
  • High solubility in DMSO (100 mg/mL) suitable for in vitro assays.
  • Formulated for small-scale research applications.
Catalog No. 50-002-22866 Supplier Medchemexpress LLC Supplier No. HY136149A5MG
Mpro inhibitor N3 hemihydrate is a small-molecule inhibitor of coronavirus main protease (Mpro) provided as a white to off-white solid hemihydrate for biochemical and antiviral research; product page lists purity, storage, and solubility information for experimental planning.
  • Potent inhibitor of coronavirus main protease with reported cellular activity.
  • High purity (99.86%).
  • Supplied as a white to off-white solid hemihydrate.
  • Stable when stored as powder at -20°C for up to 3 years.
  • High solubility in DMSO (100 mg/mL) suitable for in vitro assays.
  • Formulated for small-scale research applications.