Learn More
Cayman Chemical Mpro InhIbItr 11b 1mg
SDP
Catalog No. 502990780
Encompass_Preferred
An inhibitor of Mpro (IC50 = 0.04 µM in a TR-FRET assay); reduces viral yield in the culture supernatant of SARS-CoV-2-infected Vero E6 cells (EC50 = 0.72 µM); reduces viral RNA copy numbers in the same model from 1.85-50 µM
Catalog No. 50-299-0780 Supplier Cayman Chemical Supplier No. 313451 MG
Add to Cart
Add to Cart
An inhibitor of Mpro (IC50 = 0.04 µM in a TR-FRET assay); reduces viral yield in the culture supernatant of SARS-CoV-2-infected Vero E6 cells (EC50 = 0.72 µM); reduces viral RNA copy numbers in the same model from 1.85-50 µM