Learn More
Medchemexpress LLC HY-13994 5mg Medchemexpress, Mps1-IN-2 CAS:1228817-38-6 Purity:>98%
SDP
Catalog No. 502081547
Encompass_Preferred
Medchemexpress, HY-13994 5mg Mps1-IN-2 CAS:1228817-38-6 Mps1-IN-2 is a potent, selective and ATP-competitive dual Mps1/Plk1 inhibitor, with an IC50 and a Kd of 145 nM and 12 nM for Mps1 and a Kd of 61 nM for Plk1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1547 Supplier Medchemexpress LLC Supplier No. HY139945MG
Medchemexpress, HY-13994 5mg Mps1-IN-2 CAS:1228817-38-6 Mps1-IN-2 is a potent, selective and ATP-competitive dual Mps1/Plk1 inhibitor, with an IC50 and a Kd of 145 nM and 12 nM for Mps1 and a Kd of 61 nM for Plk1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.