Learn More
Medchemexpress LLC Mps1-IN-3 hydrochloride | 2989453-29-2 | 98.0% | 574.09 | C26H32ClN7O4S | 100MG
SDP
Supplier:  Medchemexpress LLC HY12401A100MG
Encompass_Preferred
Mps1-IN-3 hydrochloride is a potent, selective inhibitor of the Mps1 (TTK) kinase with an IC50 of approximately 50 nM. It inhibits proliferation of glioblastoma cells and sensitizes glioblastoma xenografts to vincristine in preclinical models. The compound is supplied as the hydrochloride salt in solid form for research use.
Catalog No. 50-002-35601