Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mps1-IN-3 hydrochloride | 2989453-29-2 | 98.0% | 574.09 | C26H32ClN7O4S | 10MG
SDP

Catalog No. 5000237880
Encompass_Preferred

Mps1-IN-3 hydrochloride is a potent, selective inhibitor of the Mps1 (TTK) kinase for biochemical and cellular research, with an in vitro IC50 of about 50 nM; it has been reported to inhibit glioblastoma cell proliferation and sensitize glioblastoma models to vincristine.

  • Potent, selective Mps1 kinase inhibitor (IC50 ~50 nM).
  • Reported to inhibit glioblastoma proliferation and sensitize to vincristine.
  • High purity suitable for research (approximately 98.0%).
  • Molecular weight 574.09 and formula C26H32ClN7O4S.
  • Soluble in DMSO at ≥ 20 mg/mL for in vitro use.
  • Stored sealed at -20°C; solutions stable at -80°C for long term.
  • Supplied as a solid in small research sizes.
  • Datasheet, SDS, certificate of analysis, and spectral data available.

Catalog No. 50-002-37880 Supplier Medchemexpress LLC Supplier No. HY12401A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mps1-IN-3 hydrochloride is a potent, selective inhibitor of the Mps1 (TTK) kinase for biochemical and cellular research, with an in vitro IC50 of about 50 nM; it has been reported to inhibit glioblastoma cell proliferation and sensitize glioblastoma models to vincristine.

  • Potent, selective Mps1 kinase inhibitor (IC50 ~50 nM).
  • Reported to inhibit glioblastoma proliferation and sensitize to vincristine.
  • High purity suitable for research (approximately 98.0%).
  • Molecular weight 574.09 and formula C26H32ClN7O4S.
  • Soluble in DMSO at ≥ 20 mg/mL for in vitro use.
  • Stored sealed at -20°C; solutions stable at -80°C for long term.
  • Supplied as a solid in small research sizes.
  • Datasheet, SDS, certificate of analysis, and spectral data available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.