Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mps1-IN-3 hydrochloride | 2989453-29-2 | 98.0% | 574.09 | C26H32ClN7O4S | 25MG
SDP

Catalog No. 5000235552
Encompass_Preferred

Mps1-IN-3 hydrochloride is the hydrochloride salt of a potent and selective MPS1 (TTK) kinase inhibitor (reported IC50 ≈ 50 nM) provided for research use. The compound is supplied as a solid with high purity, chemical formula C26H32ClN7O4S, and molecular weight 574.09 g/mol; it is commonly packaged in small quantities for in vitro biochemical and cell-based studies.

  • Potent MPS1 kinase inhibitor with reported IC50 ≈ 50 nM.
  • High purity solid suitable for research applications.
  • Chemical formula C26H32ClN7O4S and molecular weight 574.09 g/mol.
  • Hydrochloride salt form to aid solubility for assays.
  • Packaged in small quantities for laboratory testing and method development.

Catalog No. 50-002-35552 Supplier Medchemexpress LLC Supplier No. HY12401A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mps1-IN-3 hydrochloride is the hydrochloride salt of a potent and selective MPS1 (TTK) kinase inhibitor (reported IC50 ≈ 50 nM) provided for research use. The compound is supplied as a solid with high purity, chemical formula C26H32ClN7O4S, and molecular weight 574.09 g/mol; it is commonly packaged in small quantities for in vitro biochemical and cell-based studies.

  • Potent MPS1 kinase inhibitor with reported IC50 ≈ 50 nM.
  • High purity solid suitable for research applications.
  • Chemical formula C26H32ClN7O4S and molecular weight 574.09 g/mol.
  • Hydrochloride salt form to aid solubility for assays.
  • Packaged in small quantities for laboratory testing and method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.