Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mps1-IN-3 hydrochloride | 2989453-29-2 | 98.0% | 574.09 g/mol | C26H32ClN7O4S | 50MG
SDP

Catalog No. 5000237103
Encompass_Preferred

Mps1-IN-3 hydrochloride is a potent, selective Mps1 (TTK) kinase inhibitor (IC50 = 50 nM) supplied as a hydrochloride salt for research use. The compound is provided as a solid with high purity and suitable solubility in DMSO for in vitro and in vivo studies.

  • Molecular formula: C26H32ClN7O4S.
  • Molecular weight: 574.09 g/mol.
  • Purity: 98.0%.
  • Appearance: Light yellow to green yellow solid.
  • Solubility: DMSO ≥ 20 mg/mL.
  • Storage: Store sealed at -20°C; in solution, -80°C up to 6 months or -20°C up to 1 month.
  • Applications: Suitable for cellular assays and xenograft studies, including sensitization experiments.
  • Packaging: Available in multiple milligram-scale quantities.

Catalog No. 50-002-37103 Supplier Medchemexpress LLC Supplier No. HY12401A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mps1-IN-3 hydrochloride is a potent, selective Mps1 (TTK) kinase inhibitor (IC50 = 50 nM) supplied as a hydrochloride salt for research use. The compound is provided as a solid with high purity and suitable solubility in DMSO for in vitro and in vivo studies.

  • Molecular formula: C26H32ClN7O4S.
  • Molecular weight: 574.09 g/mol.
  • Purity: 98.0%.
  • Appearance: Light yellow to green yellow solid.
  • Solubility: DMSO ≥ 20 mg/mL.
  • Storage: Store sealed at -20°C; in solution, -80°C up to 6 months or -20°C up to 1 month.
  • Applications: Suitable for cellular assays and xenograft studies, including sensitization experiments.
  • Packaging: Available in multiple milligram-scale quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.