Learn More
Medchemexpress LLC Mps1-IN-3 hydrochloride | 2989453-29-2 | 98.0% | 574.09 g/mol | C26H32ClN7O4S | 5MG

Supplier: Medchemexpress LLC HY12401A5MG
Mps1-IN-3 hydrochloride is a potent and selective Mps1 (TTK) kinase inhibitor (IC50 ≈ 50 nM) supplied as a research-grade hydrochloride salt. It inhibits proliferation of glioblastoma cells and can sensitize glioblastoma xenografts to vincristine, making it suitable for cell biology and cancer-kinase studies. Provided as a solid with high purity for in vitro research.
- Potent Mps1 kinase inhibitor (IC50 ≈ 50 nM).
- Supplied as hydrochloride salt, solid form.
- High purity (98.0%).
- Molecular weight 574.09 g/mol; formula C26H32ClN7O4S.
- CAS 2989453-29-2.
- Available in small-scale laboratory package sizes for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.