Learn More
Cayman Chemical Mps1tK InhIbItr 10mg
SDP
Catalog No. 503015830
Encompass_Preferred
An inhibitor of Mps1/TTK (IC50 = 5.8 nM); inhibits Mps1 phosphorylation of KNL1 and increases the rate of mitosis at 100 nM; increases the number of missegregated chromosomes at 50 and 100 nM; inhibits colony formation of DLD1, HCT116, and U2OS cells (IC50s = 24.6, 20.1, and 20.6 nM, respectively)
Catalog No. 50-301-5830 Supplier Cayman Chemical Supplier No. 2555410 MG
Add to Cart
Add to Cart
An inhibitor of Mps1/TTK (IC50 = 5.8 nM); inhibits Mps1 phosphorylation of KNL1 and increases the rate of mitosis at 100 nM; increases the number of missegregated chromosomes at 50 and 100 nM; inhibits colony formation of DLD1, HCT116, and U2OS cells (IC50s = 24.6, 20.1, and 20.6 nM, respectively)