Learn More
Medchemexpress LLC MPSD control peptide (TFA) | 99.8% | 3080.76 (free base) | 1 MG
SDP
Catalog No. 5000423866
Encompass_Preferred
MPSD TFA (MARCKS-ED TFA) is the TFA salt form of MPSD, a 25-amino acid peptide derived from the myristoylated alanine-rich C-kinase substrate (MARCKS). It functions as a biological probe capable of sensing membrane curvature and recognizing phosphatidylserine, making it useful for studying membrane shape and lipid composition.
  • Derived from MARCKS effector domain sequence
  • Senses membrane curvature
  • Recognizes phosphatidylserine
  • Useful for studying membrane shape and lipid composition
  • Appears as a white to off-white solid
  • High purity (99.8%)
  • Molecular weight: 3080.76 (free base)
Catalog No. 50-004-23866 Supplier Medchemexpress LLC Supplier No. HYP10471A1MG
MPSD TFA (MARCKS-ED TFA) is the TFA salt form of MPSD, a 25-amino acid peptide derived from the myristoylated alanine-rich C-kinase substrate (MARCKS). It functions as a biological probe capable of sensing membrane curvature and recognizing phosphatidylserine, making it useful for studying membrane shape and lipid composition.
  • Derived from MARCKS effector domain sequence
  • Senses membrane curvature
  • Recognizes phosphatidylserine
  • Useful for studying membrane shape and lipid composition
  • Appears as a white to off-white solid
  • High purity (99.8%)
  • Molecular weight: 3080.76 (free base)