Learn More
Medchemexpress LLC MPSD TFA (MARCKS-ED TFA) | 99.8% | 3080.76 | 5 MG
SDP
Catalog No. 5000423785
Encompass_Preferred
MPSD TFA is the TFA salt form of MPSD, a 25-amino acid peptide derived from the effector domain sequence of the intracellular membrane protein myristoylated alanine-rich C-kinase substrate (MARCKS). It functions as a biological probe to study membrane curvature and lipid composition, specifically recognizing phosphatidylserine.
  • TFA salt form of MPSD
  • 25-amino acid peptide
  • Derived from MARCKS protein
  • Detects membrane curvature
  • Identifies phosphatidylserine
  • Useful as a biological probe
  • White to off-white solid appearance
  • Soluble in DMSO and H2O
Catalog No. 50-004-23785 Supplier Medchemexpress LLC Supplier No. HYP10471A5MG
MPSD TFA is the TFA salt form of MPSD, a 25-amino acid peptide derived from the effector domain sequence of the intracellular membrane protein myristoylated alanine-rich C-kinase substrate (MARCKS). It functions as a biological probe to study membrane curvature and lipid composition, specifically recognizing phosphatidylserine.
  • TFA salt form of MPSD
  • 25-amino acid peptide
  • Derived from MARCKS protein
  • Detects membrane curvature
  • Identifies phosphatidylserine
  • Useful as a biological probe
  • White to off-white solid appearance
  • Soluble in DMSO and H2O