Learn More
Medchemexpress LLC MPSD TFA (MARCKS-ED TFA) | 99.8% | 3080.76 (free base) | 25 MG
SDP
Catalog No. 5000426988
Encompass_Preferred
MPSD TFA is the TFA salt form of MPSD. It is a 25-amino acid peptide derived from the effector domain sequence of the intracellular membrane protein myristoylated alanine-rich C-kinase substrate (MARCKS). This peptide has the ability to sense membrane curvature and recognize phosphatidylserine. It can be used as a biological probe to investigate membrane shape and lipid composition.
  • 25-amino acid peptide based on the MARCKS effector domain.
  • Senses membrane curvature.
  • Recognizes phosphatidylserine.
  • Utilized as a biological probe to study membrane shape and lipid composition.
Catalog No. 50-004-26988 Supplier Medchemexpress LLC Supplier No. 25MGHYP10471A
MPSD TFA is the TFA salt form of MPSD. It is a 25-amino acid peptide derived from the effector domain sequence of the intracellular membrane protein myristoylated alanine-rich C-kinase substrate (MARCKS). This peptide has the ability to sense membrane curvature and recognize phosphatidylserine. It can be used as a biological probe to investigate membrane shape and lipid composition.
  • 25-amino acid peptide based on the MARCKS effector domain.
  • Senses membrane curvature.
  • Recognizes phosphatidylserine.
  • Utilized as a biological probe to study membrane shape and lipid composition.