Learn More
TARGETMOL CHEMICALS INC MPT0B014 5MG
SDP
Catalog No. 502579257
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MPT0B014 is a potent tubulin polymerization inhibitor. MPT0B014 can induce cancer cell apoptosis. purity: 99%
Catalog No. 50-257-9257 Supplier TARGETMOL CHEMICALS INC Supplier No. T677695MG
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MPT0B014 is a potent tubulin polymerization inhibitor. MPT0B014 can induce cancer cell apoptosis. purity: 99%