Learn More
Medchemexpress LLC Mpt0b390 | 1817802-18-8 | 98.8% | C17H17N3O5S | 100 MG
SDP
Catalog No. 5000355770
Encompass_Preferred
MPT0B390 is an arylsulfonamide-based derivative that exhibits potent HDAC inhibitory ability. It acts as a TIMP3 inducer and is shown to inhibit tumor growth, metastasis, and angiogenesis. This compound demonstrates antiproliferative activity against the human colon cancer cell line HCT116 with a GI50 of 0.03 μM.
  • Exhibits potent HDAC inhibitory ability
  • Acts as a TIMP3 inducer
  • Inhibits tumor growth, metastasis, and angiogenesis
  • Demonstrates antiproliferative activity against human colon cancer cell line HCT116 with a GI50 of 0.03 μM
Catalog No. 50-003-55770 Supplier Medchemexpress LLC Supplier No. HY145426100MG
MPT0B390 is an arylsulfonamide-based derivative that exhibits potent HDAC inhibitory ability. It acts as a TIMP3 inducer and is shown to inhibit tumor growth, metastasis, and angiogenesis. This compound demonstrates antiproliferative activity against the human colon cancer cell line HCT116 with a GI50 of 0.03 μM.
  • Exhibits potent HDAC inhibitory ability
  • Acts as a TIMP3 inducer
  • Inhibits tumor growth, metastasis, and angiogenesis
  • Demonstrates antiproliferative activity against human colon cancer cell line HCT116 with a GI50 of 0.03 μM