Learn More
Medchemexpress LLC Mpt0b390 | 1817802-18-8 | 98.8% | C17H17N3O5S | 10MG
SDP
Catalog No. 5000260975
Encompass_Preferred
MPT0B390 is a small-molecule arylsulfonamide that functions as a histone deacetylase (HDAC) inhibitor and induces TIMP3 expression. It demonstrates antiproliferative activity and suppresses tumor growth, metastasis, and angiogenesis in preclinical models. Supplied as an off-white to light-yellow solid for research use, the compound is provided with a reported high purity and defined storage recommendations.
  • Acts as an HDAC inhibitor and TIMP3 inducer in cellular assays.
  • Exhibits antiproliferative activity against cancer cell lines.
  • Inhibits tumor growth, metastasis, and angiogenesis in preclinical studies.
  • Available as a solid with high reported purity suitable for research use.
  • Stable under recommended storage conditions for extended shelf life.
Catalog No. 50-002-60975 Supplier Medchemexpress LLC Supplier No. HY14542610MG
MPT0B390 is a small-molecule arylsulfonamide that functions as a histone deacetylase (HDAC) inhibitor and induces TIMP3 expression. It demonstrates antiproliferative activity and suppresses tumor growth, metastasis, and angiogenesis in preclinical models. Supplied as an off-white to light-yellow solid for research use, the compound is provided with a reported high purity and defined storage recommendations.
  • Acts as an HDAC inhibitor and TIMP3 inducer in cellular assays.
  • Exhibits antiproliferative activity against cancer cell lines.
  • Inhibits tumor growth, metastasis, and angiogenesis in preclinical studies.
  • Available as a solid with high reported purity suitable for research use.
  • Stable under recommended storage conditions for extended shelf life.