Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mpt0b390 | 1817802-18-8 | 98.8% | C17H17N3O5S | 50 MG
SDP

Catalog No. 5000355333
Encompass_Preferred

MPT0B390 is an arylsulfonamide-based derivative with potent HDAC inhibitory ability. It acts as a TIMP3 inducer, inhibiting tumor growth, metastasis, and angiogenesis. This compound demonstrates antiproliferative activity against the human colon cancer cell line HCT116 with a GI50 of 0.03 μM.

  • Potent HDAC inhibitory ability
  • TIMP3 inducer
  • Inhibits tumor growth, metastasis, and angiogenesis
  • Antiproliferative activity against human colon cancer cell line HCT116
  • Molecular weight: 375.40
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Shipping at room temperature in continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility (In Vitro): DMSO at 250 mg/mL (665.96 mM)

Catalog No. 50-003-55333 Supplier Medchemexpress LLC Supplier No. HY14542650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MPT0B390 is an arylsulfonamide-based derivative with potent HDAC inhibitory ability. It acts as a TIMP3 inducer, inhibiting tumor growth, metastasis, and angiogenesis. This compound demonstrates antiproliferative activity against the human colon cancer cell line HCT116 with a GI50 of 0.03 μM.

  • Potent HDAC inhibitory ability
  • TIMP3 inducer
  • Inhibits tumor growth, metastasis, and angiogenesis
  • Antiproliferative activity against human colon cancer cell line HCT116
  • Molecular weight: 375.40
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Shipping at room temperature in continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility (In Vitro): DMSO at 250 mg/mL (665.96 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.