Learn More
Medchemexpress LLC MPT0G211 | 2151853-97-1 | 99.8% | 293.32 g/mol | C17H15N3O2 | 50MG
SDP
Catalog No. 5000236712
Encompass_Preferred
MPT0G211 is a potent, orally active, and selective histone deacetylase 6 (HDAC6) inhibitor used in preclinical research. It penetrates the blood-brain barrier and has demonstrated neuroprotective effects, reduction of tau phosphorylation, and anticancer/anti-metastatic activity in published models.
  • Potent HDAC6 inhibitor (IC50 = 0.291 nM)
  • Highly selective for HDAC6 (>1,000-fold vs other HDAC isoforms)
  • Orally active and brain-penetrant
  • Demonstrated neuroprotective effects and reduced tau phosphorylation in preclinical models
  • Shows anticancer and anti-metastatic activity in preclinical studies
  • High purity for research use (99.79%)
  • Molecular formula C17H15N3O2; molecular weight 293.32 g/mol
Catalog No. 50-002-36712 Supplier Medchemexpress LLC Supplier No. HY12397650MG
MPT0G211 is a potent, orally active, and selective histone deacetylase 6 (HDAC6) inhibitor used in preclinical research. It penetrates the blood-brain barrier and has demonstrated neuroprotective effects, reduction of tau phosphorylation, and anticancer/anti-metastatic activity in published models.
  • Potent HDAC6 inhibitor (IC50 = 0.291 nM)
  • Highly selective for HDAC6 (>1,000-fold vs other HDAC isoforms)
  • Orally active and brain-penetrant
  • Demonstrated neuroprotective effects and reduced tau phosphorylation in preclinical models
  • Shows anticancer and anti-metastatic activity in preclinical studies
  • High purity for research use (99.79%)
  • Molecular formula C17H15N3O2; molecular weight 293.32 g/mol