Learn More
Bio Rad Laboratories MPTGX 12% 10 WELLS 50UL 10/PK
Catalog No. NC0038021
Encompass
Mini-PROTEAN TGX extended shelf-life gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 8.6 cm x 7.2 cm x 1.0 mm (W x L x thickness) are supplied in a 10.0 x 8.5 cm cassette.
Catalog No. NC0038021 Supplier Bio Rad Laboratories Supplier No. 4561044
Mini-PROTEAN TGX extended shelf-life gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 8.6 cm x 7.2 cm x 1.0 mm (W x L x thickness) are supplied in a 10.0 x 8.5 cm cassette.