Learn More
Medchemexpress LLC HY-15608 100mg , MPTP (hydrochloride) CAS:23007-85-4 Purity:98%
SDP
Catalog No. 501492691
Encompass_Preferred
Medchemexpress, HY-15608 100mg MPTP (hydrochloride) CAS:23007-85-4 Formula:C12H16ClN Purity:98% MPTP (hydrochloride) is a prodrug to the neurotoxin MPP + which causes permanent symptoms of Parkinson's disease. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2691 Supplier Medchemexpress LLC Supplier No. HY15608100MG
Medchemexpress, HY-15608 100mg MPTP (hydrochloride) CAS:23007-85-4 Formula:C12H16ClN Purity:98% MPTP (hydrochloride) is a prodrug to the neurotoxin MPP + which causes permanent symptoms of Parkinson's disease. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.