Learn More
Selleck Chemical LLC MPTP hydrochloride S4732-200mg
SDP
Catalog No. 503109410
Encompass_Preferred
MPTP hydrochloride is a dopaminergic neurotoxin and cause selective destruction of dopaminergic neurons in animal models of parkinsonism MPTP hydrochloride induces apoptosis MPTP hydrochloride can be used to induce animal models of Parkinson s disease
Catalog No. 50-310-9410 Supplier Selleck Chemical LLC Supplier No. S4732200MG
MPTP hydrochloride is a dopaminergic neurotoxin and cause selective destruction of dopaminergic neurons in animal models of parkinsonism MPTP hydrochloride induces apoptosis MPTP hydrochloride can be used to induce animal models of Parkinson s disease