Learn More
TARGETMOL CHEMICALS INC MPTP HYDROCHLORIDE 50MG
SDP
Catalog No. 502579969
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MPTP hydrochloride (MPTP-hydrochloride) is a precusor of MPP+ which is toxic to dopaminergic neurons and causes Parkinsonism. purity: 99%
Catalog No. 50-257-9969 Supplier TARGETMOL CHEMICALS INC Supplier No. T408150MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MPTP hydrochloride (MPTP-hydrochloride) is a precusor of MPP+ which is toxic to dopaminergic neurons and causes Parkinsonism. purity: 99%