Learn More
AdipoGen MPTS
SDP
Catalog No. 502560550
Encompass_Preferred
Chemical. CAS 82962-86-5. Formula C17H9Na3O10S3. MW 538.41. Synthetic. Highly water-soluble superpolar fluorescent probe for assaying cationic surfactants. Can be used as a polar tracer. It has also been used in combination with a boronic acid-modified viologen quencher to sense glucose at pH 7.4 in buffered aqueous solution.
Catalog No. 50-256-0550 Supplier AdipoGen Supplier No. CDXM0055M100
Chemical. CAS 82962-86-5. Formula C17H9Na3O10S3. MW 538.41. Synthetic. Highly water-soluble superpolar fluorescent probe for assaying cationic surfactants. Can be used as a polar tracer. It has also been used in combination with a boronic acid-modified viologen quencher to sense glucose at pH 7.4 in buffered aqueous solution.