Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ Blood DNA Extraction Kit 1200

Catalog No. 117022200 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
48 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-702-2200 48 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-702-2200 Supplier MP Biomedicals™ Supplier No. 117022200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For extraction and purification of genomic DNA from 400 to 1000μL mammalian whole blood or a suspension of mammalian blood cells

  • For extraction of genomic DNA, mitochondrial and viral DNA
  • Compatible with ammalian fresh or frozen whole blood
  • Completely automated purification protocol
  • Runs in as little as 60 minutes
  • Produces nucleic acids that can be used immediately in downstream applications such as PCR, qPCR, Sequencing (NGS), Microarray, RFLP and Southern Blot Analysis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description Blood DNA Extraction Kit 1200
Format Kit
Isolation Technology Magnetic Bead
Sample Type Buffy Coat, Whole Blood Cells
For Use With (Equipment) MPure-12 Automated Nucleic Acid Purification System
For Use With (Application) DNA Purification
Includes Proteinase K solution, 50μL; Lysis Buffer 2, 1500μL; Binding Buffer 1, 1000μL; Magnetic Bead Solution, 1000μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
No. of Reactions 48 Preps
Product Type DNA Extraction Kit
Sample Volume 400 to 1000μL whole blood (WBC count less than 6 x 104 cells/μL); 400 to 1000μL buffy coat (contains no more than 5 x 106 cells)
Test Time As little as 60 minutes
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.