Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ Blood DNA Extraction Kit 200

Catalog No. MP117022100 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
48 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP117022100 48 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP117022100 Supplier MP Biomedicals™ Supplier No. 117022100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For extraction of DNA from mammalian whole blood or a suspension of mammalian blood cells

MPure Blood DNA Extraction Kit 200 is used with the MPure-12 aNAP System for extraction of DNA from 100-400μL mammalian whole blood or suspension of mammalian blood cells. Nucleic acids extracted from MPure Blood DNA Extraction Kit 200 can be used in a number of downstream application including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP, and Southern Blot Analysis.

  • For extraction of host gDNA, mitochondrial and viral DNA
  • Extracts DNA from 100-400µL mammalian whole blood
  • Nucleic acids can be used immediately in downstream applications
  • Completely automated
  • Can be run in as little as 50 minutes

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description Blood DNA Extraction Kit 200
Format Kit
Isolation Technology Magnetic Bead
Sample Type Buffy Coat, Leukocytes, Whole Blood Cells
For Use With (Equipment) MPure-12 Automated Nucleic Acid Purification System
For Use With (Application) Nucleic Acid Purification
Includes Proteinase K solution, 40μL; Lysis Buffer 2, 1000μL; Binding Buffer 1, 600μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
No. of Reactions 48 Preps
Product Type DNA Extraction Kit
Sample Volume 100-400μL whole blood (WBC count less than 2 x 104 cells/μL); 100-400μL leukocyte concentration (contain no more than 5 x 106 cells) ; 100-400μL buffy coat
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.