Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ FFPE DNA Extraction Kit

Catalog No. 117022900 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
48 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-702-2900 48 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-702-2900 Supplier MP Biomedicals™ Supplier No. 117022900

For the extraction and purification of genomic DNA from FFPE (Formalin-Fixed, Paraffin-Embedded) tissue samples

  • Completely automated purification protocol
  • Run times as little as 35 minutes
  • DNA can be used immediately in downstream applications

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description FFPE DNA Extraction Kit
Isolation Technology Magnetic Bead
Sample Type FFPE
For Use With (Equipment) MPure-12 Automated Nucleic Acid Purification System
For Use With (Application) DNA Purification
Includes Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
No. of Reactions 48 Preps
Sample Volume 100 - 400μL; One to eight 10 μm- thick sections of tissue; Three to ten biopsies.
Test Time As little as 35 minutes
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.