Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ Forensic DNA Extraction Kit

Catalog No. 117022110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
48 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-702-2110 48 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-702-2110 Supplier MP Biomedicals™ Supplier No. 117022110

missing translation for 'validMainframeMsg'

Extract and isolate genomic DNA from forensic samples

The kit contains everything needed for sample preparation. However, for samples such as swipes and swabs, cigarette butts and bodily fluid stained fabrics or other solid matrices, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing time. DNA extracted using the MPure Forensic DNA Extraction Kit is ready for immediate use in downstream applications.

  • Completely automated purification protocol
  • Run in as little as 40 minutes
  • DNA is ready for immediate use

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description Forensic DNA Extraction Kit
Sample Type Forensic Sample
For Use With (Equipment) MPure-12™ Automated Nucleic Acid Purification System
For Use With (Application) Nucleic Acid Purification; Genomic DNA from Forensic samples
Includes Lysis Buffer 2, 1000μL; Binding Buffer 1, 1000μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
No. of Reactions 48 Preps
Sample Volume Clotted/ dried blood: 20μL; Forensic Surface and Contact Swabs, Hair root: 0.5 to 1cm; Human tissues: 40mg; Saliva: 50μL; Sperm Stains: 5 to 10μL or 1cm2 of the forensic sample; Chewing Gum: 40 mg; Cigarette butts: 1cm2 paper from the end of the cigarette or filter
Test Time As little as 40 minutes
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.