Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ Plant DNA Extraction Kit

Catalog No. 117022150 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
48 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-702-2150 48 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-702-2150 Supplier MP Biomedicals™ Supplier No. 117022150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL

For extraction of genomic DNA from plant and fungal tissues

MPure Plant DNA Extraction Kit contains everything needed for sample preparation. However, for tough tissues such as roots and seeds, or tough fungal tissue, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can increase yields as well as shorten processing time.

  • Compatible with fresh or lyophilized plant tissue as well as fully formed mycelium or cells from yeast culture
  • Completely automated purification protocol
  • Runs in as little as 40 minutes
  • Generates DNA that can be used immediately in downstream applications

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description Plant DNA Extraction Kit
Format Kit
Isolation Technology Magnetic Bead
For Use With (Equipment) MPure-12 Automated Nucleic Acid Purification System
For Use With (Application) DNA Purification
Includes Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
No. of Reactions 48 Preps
Product Type DNA Extraction Kit
Test Time As little as 40 minutes
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.