Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ TB DNA Extraction Kit

Catalog No. MP117022800 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
48 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP117022800 48 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP117022800 Supplier MP Biomedicals™ Supplier No. 117022800

missing translation for 'validMainframeMsg'

For extraction of genomic DNA of Mycobacteria spp. (e.g. Mycobacterium tuberculosis) from different specimens

  • Variety of biological samples including sputum, bronchoalveolar lavage, pus, blood, cell-free body fluids, urine and respiratory specimens
  • Solid or suspended bacterial culture
  • Completely automated purification protocol
  • Run times as little as 60 minutes
  • DNA can be used immediately in downstream applications

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description TB DNA Extraction Kit
Isolation Technology Magnetic Bead
Sample Type Mycobacterium Specimens
For Use With (Equipment) MPure-12 Automated Nucleic Acid Purification System
For Use With (Application) DNA Purification
Includes Proteinase K solution, 40μL; Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL; Buffer N1, 400μL
No. of Reactions 48 Preps
Test Time As little as 60 minutes
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.