Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ Viral Nucleic Acid Extraction Kit

Catalog No. 117022300 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
48 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-702-2300 48 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-702-2300 Supplier MP Biomedicals™ Supplier No. 117022300
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For extraction of Viral DNA or RNA from human biological specimens such as serum, plasma, CSF, urine and other cell-free fluids

  • Completely automated purification protocol
  • Run in as little as 40 minutes
  • Nucleic Acids can be used immediately in downstream applications such as PCR, qPCR, Sequencing (NGS), Microarray, RFLP, and Southern Blot Analysis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description Viral Nucleic Acid Extraction Kit
Format Kit
Isolation Technology Magnetic Bead
Sample Type Human Biological Specimens (Serum, Plasma, and Other Cell-free Fluids)
For Use With (Equipment) MPure-12 Automated Nucleic Acid Purification System
For Use With (Application) Viral DNA or RNA Purification
Includes Proteinase K solution, 30μL; Lysis Buffer 2, 720μL; Binding Buffer 1, 1000μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL: RNase-free water, 1000μL; RNase-free water, 1000μL
No. of Reactions 48 Preps
Product Type Nucleic Acid Extraction Kit
Sample Volume 100 to 400μL
Test Time As little as 40 minutes
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.