Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ Viral Nucleic Acid Extraction Kit 800

Catalog No. 117022170 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
48 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-702-2170 48 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-702-2170 Supplier MP Biomedicals™ Supplier No. 117022170

missing translation for 'validMainframeMsg'

For extraction of Viral DNA or RNA from human biological specimens such as serum, plasma, and other cell-free fluids

  • Used with the MPure-12 instrument for extraction
  • Completely automated purification protocol
  • Run in as little as 55 minutes
  • Nucleic acids can be used immediately in downstream applications

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description Viral Nucleic Acid Extraction Kit 800
Sample Type Human Biological Specimens (Serum, Plasma, and Other Cell-free Fluids)
For Use With (Equipment) MPure-12 Automated Nucleic Acid Purification System
For Use With (Application) Nucleic Acid Purification; Viral DNA or RNA from human biological specimens such as serum, plasma, and other cell-free fluids
Includes Proteinase K Solution: 40μL; Lysis Buffer 2: 1000μL; Binding Buffer 1: 1600μL; Magnetic Bead Solution: 800μL; Washing Buffer 1: 1000μL; Washing Buffer 2: 1000μL; Washing Buffer 3: 1000μL; RNase Free Water: 1000μL
No. of Reactions 48 Preps
Sample Volume 600 to 800μL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.