Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ Viral/Pathogen Nucleic Acids Extraction Kit B

Catalog No. 117022130 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
48 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-702-2130 48 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-702-2130 Supplier MP Biomedicals™ Supplier No. 117022130
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For bacterial culture, swab samples and environmental samples

MPure Viral/Pathogen Nucleic Acids Extraction Kit B is used with the MPure-12 instrument for extraction of viral and bacterial DNA/RNA from bacterial culture, swab samples and environmental samples. Starting samples include clinical specimens such as sputum, bronchoalveolar lavage, cell-free body fluids, urine, respiratory specimens, bacterial culture either solid or in suspension and environmental samples such as soil, water and feces. The kit contains everything needed for sample preparation. However, for tough samples (soil or feces), sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing times.

  • Completely automated purification protocol
  • Can be run in as little as 55 minutes
  • Extracted nucleic acids can be used in downstream applications

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description Viral/Pathogen Nucleic Acids Extraction Kit B
Format Kit
Isolation Technology Magnetic Bead
Sample Type Bacteria, Virus, Swab
For Use With (Equipment) MPure-12 Automated Nucleic Acid Purification System
For Use With (Application) Nucleic Acid Purification; viral and bacterial DNA/RNA from cell-rich samples
Includes Proteinase K: 40μL; Lysis Buffer 2: 720μL; Binding Buffer 1: 720μL; Magnetic Bead Solution: 800μL; Washing Buffer 1: 1000μL; Washing Buffer 2: 1000μL; Washing Buffer 3: 1000μL; Elution Buffer 1: 1000μL; Elution Buffer 2: 1000μL; BL2 Buffer: 400μL
No. of Reactions 48 Preps
Product Type Nucleic Acid Extraction Kit
Sample Volume Bacteria Pellet: 100-200μL /Up to 109 bacteria (about OD600 = 3); Bacterial Colony: 100-200μL /1-3 colony; Swab samples: 100-200μL liquid transport media
Test Time As little as 55 minutes
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.