Learn More
Aviva Systems Biology Corp MPV17L Antibody - N-terminal region (ARP55695_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55695_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MPV17L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MPV17L. Alternate names: FLJ39599, MGC70356, MLPH1, MLPH2, M-LPH, MPV17L1ApplicationWB. Species reactivity: Guinea Pig, Human
Catalog No. 50-014-61583