Learn More
Sino Biological MPXV MPXV-A29L MAb 100ug (Sino Biological)
Supplier:  Sino Biological 40891M0031H100
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-237-2865