Learn More
Sino Biological MPXV MPXV-A29L MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40891MM2420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Monkeypox Virus (MPXV) Protein A29 (Catalog#40891-V08E). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2281