Learn More
Sino Biological MPXV MPXV-A5L PAb 100ul (Sino Biological)
Supplier:  Sino Biological 40905T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Monkeypox Virus (MPXV) A5L Protein ( Catalog#40905-V07E). Monkeypox Virus (MPXV) A5L specific IgG was purified by Monkeypox Virus (MPXV) A5L affinity chromatography.
Catalog No. 50-237-3286