Learn More
Sino Biological MPXV MPXV-B6R Protein 20ug (Sino Biological)
Supplier:  Sino Biological 40902V08HB20
Encompass_Preferred
A DNA sequence encoding the Monkeypox Virus (MPXV) B6R Protein was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-237-1563