Learn More
Sino Biological Monkeypox Virus (MPXV) A29 Antibody (HRP), Mouse MAb
Supplier:  Sino Biological 40891M0031H20
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-3584