Learn More
Sino Biological Monkeypox Virus (MPXV) (Clade Ib) Protein A35R (His Tag)
Catalog No. 502993344
Encompass_Preferred
A DNA sequence encoding the Monkeypox Virus (MPXV) A35R Protein (EPI_ISL_19093796) (Arg58-Thr181) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-299-3344 Supplier Sino Biological Supplier No. 41043V07E100
A DNA sequence encoding the Monkeypox Virus (MPXV) A35R Protein (EPI_ISL_19093796) (Arg58-Thr181) was expressed with a polyhistidine tag at the N-terminus.